Assay Details
| Gene Symbol: CDHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008231.4 | XP_004008280.3 | 14 - 15 | 2190 | 66 | |
| XM_027968585.1 | XP_027824386.1 | 13 - 14 | 2091 | 66 | ||
More Information
Set Membership: |
|

CDHR3
Genomic Map