Assay Details
| Gene Symbol: CENATAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979282.1 | XP_027835083.1 | 9 - 10 | 908 | 107 | |
| XM_027979283.1 | XP_027835084.1 | 9 - 10 | 854 | 107 | ||
More Information
Set Membership: |
|

CENATAC
Genomic Map