Assay Details
| Gene Symbol: PAK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019137.3 | XP_004019186.1 | 3 - 4 | 440 | 67 | |
| XM_012101305.2 | XP_011956695.1 | 3 - 4 | 443 | 67 | ||
More Information
Set Membership: |
|

PAK1IP1
Genomic Map