Assay Details
| Gene Symbol: TIMM10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096449.3 | XP_011951839.1 | 2 - 3 | 172 | 115 | |
| XM_012096450.3 | XP_011951840.1 | 2 - 3 | 200 | 115 | ||
More Information
Set Membership: |
|

TIMM10
Genomic Map