Assay Details
| Gene Symbol: C20H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958868.1 | XP_027814669.1 | 4 - 5 | 1552 | 91 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C20H6orf136| Gene Symbol: C20H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958868.1 | XP_027814669.1 | 4 - 5 | 1552 | 91 | |