Assay Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106577.3 | XP_011961967.1 | 11 - 12 | 1442 | 70 | |
XM_012106579.3 | XP_011961969.1 | 11 - 12 | 1442 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
