Assay Details
| Gene Symbol: LOC101107282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970762.1 | XP_027826563.1 | 49 - 50 | 8513 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101107282| Gene Symbol: LOC101107282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970762.1 | XP_027826563.1 | 49 - 50 | 8513 | 92 | |