Assay Details
| Gene Symbol: STIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970914.1 | XP_027826715.1 | 7 - 8 | 1414 | 81 | |
| XM_027970915.1 | XP_027826716.1 | 7 - 8 | 1414 | 81 | ||
More Information
Set Membership: |
|

STIM2
Genomic Map