Assay Details
| Gene Symbol: LOC101109892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186372.3 | XP_012041762.1 | 6 - 7 | 1035 | 64 | |
| XM_027974315.1 | XP_027830116.1 | 6 - 7 | 583 | 64 | ||
| XM_027974316.1 | XP_027830117.1 | 6 - 7 | 574 | 64 | ||
More Information
Set Membership: |
|

LOC101109892
Genomic Map