Assay Details
| Gene Symbol: C9H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973442.1 | XP_027829243.1 | 3 - 4 | 343 | 82 | |
| XM_027973443.1 | XP_027829244.1 | 4 - 5 | 930 | 82 | ||
More Information
Set Membership: |
|

C9H8orf33
Genomic Map