Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101418.3 | XP_011956808.1 | 3 - 4 | 496 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR87| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101418.3 | XP_011956808.1 | 3 - 4 | 496 | 98 | |
Set Membership: |
|