Assay Details
Gene Symbol: TAFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967497.1 | XP_027823298.1 | 1 - 2 | 116 | 120 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
