Assay Details
| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012229.3 | XP_004012278.2 | 7 - 8 | 1619 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

POGLUT2| Gene Symbol: POGLUT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012229.3 | XP_004012278.2 | 7 - 8 | 1619 | 81 | |