Assay Details
| Gene Symbol: TRIM68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016261.3 | XP_004016310.1 | 3 - 4 | 653 | 66 | |
| XM_012095952.3 | XP_011951342.2 | 2 - 3 | 165 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_012095952.3 transcripts.
Set Membership: |
|

TRIM68
Genomic Map