Assay Details
| Gene Symbol: HTATSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015105010.2 | XP_014960496.1 | 8 - 9 | 1192 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTATSF1| Gene Symbol: HTATSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015105010.2 | XP_014960496.1 | 8 - 9 | 1192 | 68 | |
Set Membership: |
|