Assay Details
| Gene Symbol: LOC101113672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968391.1 | XP_027824192.1 | 31 - 32 | 3694 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113672| Gene Symbol: LOC101113672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968391.1 | XP_027824192.1 | 31 - 32 | 3694 | 111 | |
Set Membership: |
|