Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968014.1 | XP_027823815.1 | 1 - 2 | 156 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

WASHC1| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968014.1 | XP_027823815.1 | 1 - 2 | 156 | 93 | |