Assay Details
| Gene Symbol: LOC101111247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104097.2 | XP_014959583.2 | 24 - 25 | 2929 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111247| Gene Symbol: LOC101111247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104097.2 | XP_014959583.2 | 24 - 25 | 2929 | 83 | |