Assay Details
| Gene Symbol: ZNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978938.1 | XP_027834739.1 | 3 - 4 | 354 | 102 | |
| XM_027978939.1 | XP_027834740.1 | 3 - 4 | 354 | 102 | ||
More Information
Set Membership: |
|

ZNF8
Genomic Map