Assay Details
| Gene Symbol: PDE6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094086.2 | XP_014949572.1 | 1 - 2 | 168 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDE6H| Gene Symbol: PDE6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094086.2 | XP_014949572.1 | 1 - 2 | 168 | 102 | |
Set Membership: |
|