Assay Details
| Gene Symbol: GPATCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012987.4 | XP_004013036.3 | 3 - 4 | 372 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPATCH8| Gene Symbol: GPATCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012987.4 | XP_004013036.3 | 3 - 4 | 372 | 60 | |
Set Membership: |
|