Assay Details
| Gene Symbol: LOC101117600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013284.4 | XP_004013333.1 | 6 - 7 | 723 | 66 | |
| XM_027975008.1 | XP_027830809.1 | 5 - 6 | 628 | 66 | ||
| XM_027975009.1 | XP_027830810.1 | 5 - 6 | 609 | 66 | ||
More Information
Set Membership: |
|

LOC101117600
Genomic Map