Assay Details
| Gene Symbol: MRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960813.1 | XP_027816614.1 | 5 - 6 | 737 | 65 | |
| XM_027960814.1 | XP_027816615.1 | 5 - 6 | 737 | 65 | ||
More Information
Set Membership: |
|

MRO
Genomic Map