Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103836.3 | XP_011959226.2 | 5 - 6 | 611 | 93 | |
| XM_012103839.2 | XP_011959229.2 | 4 - 5 | 377 | 93 | ||
More Information
Set Membership: |
|

SERPINB13
Genomic Map