Assay Details
| Gene Symbol: ZNF76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959159.1 | XP_027814960.1 | 5 - 6 | 785 | 71 | |
| XM_027959160.1 | XP_027814961.1 | 5 - 6 | 660 | 71 | ||
| XM_027959161.1 | XP_027814962.1 | 4 - 5 | 669 | 71 | ||
| XM_027959162.1 | XP_027814963.1 | 5 - 6 | 747 | 71 | ||
| XM_027959163.1 | XP_027814964.1 | 5 - 6 | 857 | 71 | ||
| XM_027959165.1 | XP_027814966.1 | 5 - 6 | 747 | 71 | ||
More Information
Set Membership: |
|

ZNF76
Genomic Map