Assay Details
| Gene Symbol: SLC22A20P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959621.1 | XP_027815422.1 | 8 - 9 | 1370 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A20P| Gene Symbol: SLC22A20P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959621.1 | XP_027815422.1 | 8 - 9 | 1370 | 62 | |
Set Membership: |
|