Assay Details
| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971745.1 | XP_027827546.1 | 7 - 8 | 692 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTRAF| Gene Symbol: RTRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971745.1 | XP_027827546.1 | 7 - 8 | 692 | 96 | |