Assay Details
| Gene Symbol: LOC101115706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969603.1 | XP_027825404.1 | 19 - 20 | 2517 | 66 | |
| XM_027969604.1 | XP_027825405.1 | 18 - 19 | 2291 | 66 | ||
More Information
Set Membership: |
|

LOC101115706
Genomic Map