Assay Details
| Gene Symbol: TNFRSF11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960852.1 | XP_027816653.1 | 6 - 7 | 617 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF11A| Gene Symbol: TNFRSF11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960852.1 | XP_027816653.1 | 6 - 7 | 617 | 62 | |
Set Membership: |
|