Assay Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098563.2 | XP_014954049.2 | 1 - 2 | 350 | 102 | |
| XM_027975151.1 | XP_027830952.1 | 1 - 2 | 353 | 102 | ||
More Information
Set Membership: |
|

CRK
Genomic Map