Assay Details
| Gene Symbol: HOPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180015.3 | XP_012035405.2 | 2 - 3 | 554 | 77 | |
| XM_012180016.3 | XP_012035406.2 | 2 - 3 | 259 | 77 | ||
| XM_027971019.1 | XP_027826820.1 | 2 - 3 | 265 | 77 | ||
| XM_027971020.1 | XP_027826821.1 | 2 - 3 | 256 | 77 | ||
| XM_027971021.1 | XP_027826822.1 | 2 - 3 | 530 | 77 | ||
More Information
Set Membership: |
|

HOPX
Genomic Map