Assay Details
| Gene Symbol: LOC101122080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093334.2 | XP_014948820.1 | 15 - 16 | 1759 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122080| Gene Symbol: LOC101122080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093334.2 | XP_014948820.1 | 15 - 16 | 1759 | 89 | |