Assay Details
| Gene Symbol: LOC114113113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964796.1 | XP_027820597.1 | 1 - 2 | 144 | 162 | |
| Gene Symbol: LOC101122080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093334.2 | XP_014948820.1 | 6 - 7 | 589 | 162 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027964796.1 transcripts.
Set Membership: |
|

LOC114113113
Genomic Map