Assay Details
| Gene Symbol: CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103123.2 | XP_014958609.2 | 3 - 4 | 642 | 56 | |
| XM_027959389.1 | XP_027815190.1 | 3 - 4 | 642 | 56 | ||
| XM_027959390.1 | XP_027815191.1 | 3 - 4 | 642 | 56 | ||
| XM_027959391.1 | XP_027815192.1 | 3 - 4 | 642 | 56 | ||
| XM_027959392.1 | XP_027815193.1 | 3 - 4 | 642 | 56 | ||
| XM_027959393.1 | XP_027815194.1 | 3 - 4 | 642 | 56 | ||
More Information
Set Membership: |
|

CD6
Genomic Map