Assay Details
| Gene Symbol: AHSA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094330.2 | XP_014949816.2 | 2 - 3 | 363 | 105 | |
| XM_015094331.2 | XP_014949817.2 | 2 - 3 | 363 | 105 | ||
More Information
Set Membership: |
|

AHSA2P
Genomic Map