Assay Details
| Gene Symbol: GPIHBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183471.3 | XP_012038861.2 | 3 - 4 | 386 | 96 | |
| XM_027973445.1 | XP_027829246.1 | 2 - 3 | 877 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPIHBP1| Gene Symbol: GPIHBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012183471.3 | XP_012038861.2 | 3 - 4 | 386 | 96 | |
| XM_027973445.1 | XP_027829246.1 | 2 - 3 | 877 | 96 | ||