Assay Details
| Gene Symbol: LOC101122744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979022.1 | XP_027834823.1 | 8 - 9 | 2418 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122744| Gene Symbol: LOC101122744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979022.1 | XP_027834823.1 | 8 - 9 | 2418 | 79 | |
Set Membership: |
|