Assay Details
Gene Symbol: LOC101111607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004928.4 | XP_004004977.1 | 5 - 6 | 999 | 102 | |
XM_012146300.3 | XP_012001690.1 | 5 - 6 | 916 | 102 | ||
XM_012146310.3 | XP_012001700.1 | 5 - 6 | 1066 | 102 | ||
XM_027965189.1 | XP_027820990.1 | 5 - 6 | 843 | 102 | ||
XM_027965190.1 | XP_027820991.1 | 5 - 6 | 946 | 102 | ||
XM_027965192.1 | XP_027820993.1 | 5 - 6 | 810 | 102 |

More Information
Set Membership: |
![]() ![]() |
