Assay Details
| Gene Symbol: TM4SF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096360.3 | XP_011951750.2 | 3 - 4 | 445 | 69 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM4SF19| Gene Symbol: TM4SF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096360.3 | XP_011951750.2 | 3 - 4 | 445 | 69 | |