Assay Details
| Gene Symbol: LOC101122753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104525.3 | XP_011959915.3 | 9370 | 63 | ||
| XM_027961385.1 | XP_027817186.1 | 9369 | 63 | |||
| XM_027961387.1 | XP_027817188.1 | 9105 | 63 | |||
| XM_027961388.1 | XP_027817189.1 | 9057 | 63 | |||
More Information
Set Membership: |
|

LOC101122753
Genomic Map