Assay Details
| Gene Symbol: CTIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103729.2 | XP_014959215.2 | 10 - 11 | 1605 | 67 | |
| XM_027960717.1 | XP_027816518.1 | 10 - 11 | 1611 | 67 | ||
More Information
Set Membership: |
|

CTIF
Genomic Map