Assay Details
| Gene Symbol: SRPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962914.1 | XP_027818715.1 | 3 - 4 | 438 | 60 | |
| XM_027962915.1 | XP_027818716.1 | 3 - 4 | 306 | 60 | ||
| XM_027962916.1 | XP_027818717.1 | 2 - 3 | 377 | 60 | ||
More Information
Set Membership: |
|

SRPX
Genomic Map