Assay Details
| Gene Symbol: KCNA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012175539.3 | XP_012030929.2 | 1 - 2 | 5284 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNA6| Gene Symbol: KCNA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012175539.3 | XP_012030929.2 | 1 - 2 | 5284 | 74 | |
Set Membership: |
|