Assay Details
| Gene Symbol: SIAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092602.2 | XP_014948088.2 | 1 - 2 | 917 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIAH2| Gene Symbol: SIAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092602.2 | XP_014948088.2 | 1 - 2 | 917 | 102 | |
Set Membership: |
|