Assay Details
| Gene Symbol: LOC114108867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956952.1 | XP_027812753.1 | 2 - 3 | 2559 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114108867| Gene Symbol: LOC114108867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956952.1 | XP_027812753.1 | 2 - 3 | 2559 | 91 | |
Set Membership: |
|