Assay Details
| Gene Symbol: LOC114108867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956952.1 | XP_027812753.1 | 3 - 4 | 3586 | 141 | |
| Gene Symbol: DCHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956223.1 | XP_027812024.1 | 5 - 6 | 4068 | 141 | |
More Information
Set Membership: |
|

LOC114108867
Genomic Map