Assay Details
| Gene Symbol: KRT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974529.1 | XP_027830330.1 | 1 - 2 | 450 | 71 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT19| Gene Symbol: KRT19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974529.1 | XP_027830330.1 | 1 - 2 | 450 | 71 | |