Assay Details
| Gene Symbol: FZD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097887.2 | XP_014953373.2 | 3 - 4 | 659 | 65 | |
| XM_015097888.2 | XP_014953374.2 | 3 - 4 | 634 | 65 | ||
More Information
Set Membership: |
|

FZD6
Genomic Map