Assay Details
| Gene Symbol: SLC25A37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964323.1 | XP_027820124.1 | 2 - 3 | 1350 | 78 | |
| XM_027964324.1 | XP_027820125.1 | 2 - 3 | 420 | 78 | ||
More Information
Set Membership: |
|

SLC25A37
Genomic Map