Assay Details
| Gene Symbol: GALNT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010747.3 | XP_004010796.2 | 3 - 4 | 807 | 66 | |
| XM_027971949.1 | XP_027827750.1 | 4 - 5 | 704 | 66 | ||
| XM_027971950.1 | XP_027827751.1 | 3 - 4 | 807 | 66 | ||
More Information
Set Membership: |
|

GALNT16
Genomic Map